Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
500 g
1 kg
Catalog number ACR404160025
Price (MYR)
1,004.00
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (MYR)
1,004.00
Add to cart
Specifications
Chemical Name or MaterialCastor oil
CAS8001-79-4
Melting Point-10°C
ColorYellow
Density0.9570g/mL