5(6)-Carboxyfluorescein
5(6)-Carboxyfluorescein
5(6)-Carboxyfluorescein
5(6)-Carboxyfluorescein
Thermo Scientific Chemicals
5(6)-Carboxyfluorescein
CAS: 72088-94-9 | C42H24O14 | 752.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
Packaging:
Glass bottle
Catalog number ACR404105000
Price (MYR)
455.00
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
455.00
Add to cart
Chemical Identifiers
CAS12036-44-1
IUPAC Namedithulium(3+) trioxidandiide
Molecular FormulaO3Tm2
InChI KeyZIKATJAYWZUJPY-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Tm+3].[Tm+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.003% max.