Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
Packaging:
Glass bottle
Catalog number ACR404045000
Price (MYR)
603.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
603.00
Add to cart
Specifications
Chemical Name or MaterialActivated charcoal,NORIT* CN 1
Name NoteNORIT CN 1
CAS7440-44-0
Health Hazard 1GHS Signal Word: Warning
Melting Point3652.0°C
View more
Filter aid
RUO – Research Use Only