Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Packaging:
Glass bottle
Catalog number ACR404042500
Price (MYR)
438.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
438.00
Add to cart
Chemical Identifiers
CAS56-40-6
IUPAC Name2-aminoacetic acid
Molecular FormulaC2H5NO2
InChI KeyDHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILESNCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=99.0 % (on dried substance)
Residual solventsMeets the requirements
Arsenic (As)=<1 ppm
Infrared spectrumConforms
Loss on drying=<0.2 % (105°C, 2 h)
View more
Filter aid
RUO – Research Use Only