Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
CAS: 5794-28-5 | C2H2CaO5 | 146.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
Packaging:
Glass bottle
Catalog number ACR403885000
Price (MYR)
732.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
732.00
Add to cart
Chemical Identifiers
CAS67-64-1
IUPAC Namepropan-2-one
Molecular FormulaC3H6O
InChI KeyCSCPPACGZOOCGX-UHFFFAOYSA-N
SMILESCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(in water) Passes test
Color scale=<10 APHA
GC>=99.5 %
Residue after evaporation=<0.001 %
Titrable acid=<0.0003 meq/g