Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
CAS: 5794-28-5 | C2H2CaO5 | 146.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
Packaging:
Glass bottle
Catalog number ACR403880050
Price (MYR)
394.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
394.00
Add to cart
Chemical Identifiers
CAS622-97-9
IUPAC Name1-ethenyl-4-methylbenzene
Molecular FormulaC9H10
InChI KeyJLBJTVDPSNHSKJ-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
Refractive index1.5410 to 1.5430 (20°C, 589 nm)