Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number ACR403575000
Price (MYR)
1,248.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,248.00
Add to cart
Chemical Identifiers
CAS14694-95-2
IUPAC Nameλ¹-rhodium(1+) tris(triphenylphosphane) chloride
Molecular FormulaC54H45ClP3Rh
InChI KeyIXAYKDDZKIZSPV-UHFFFAOYSA-M
SMILES[Cl-].[Rh+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<0.01 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)Brown