Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number ACR403571000
Price (MYR)
713.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
713.00
Add to cart
Specifications
Chemical Name or MaterialButyl 4-hydroxybenzoate
Name Note99+%
CAS94-26-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement