2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
CAS: 20769-85-1 | C4H6Br2O | 229.9 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR403095000
Price (MYR)
1,783.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,783.00
Add to cart
Specifications
Chemical Name or Material2-Bromo-2-methylpropionyl bromide
CAS20769-85-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0