2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
CAS: 20769-85-1 | C4H6Br2O | 229.9 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR403091000
Price (MYR)
576.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
576.00
Add to cart
Chemical Identifiers
CAS69-52-3
IUPAC Namesodium (2S,5R,6R)-6-[(2R)-2-amino-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular FormulaC16H18N3NaO4S
InChI KeyKLOHDWPABZXLGI-YWUHCJSESA-M
SMILES[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystals or crystalline powder
Assay from Suppliers CofAPotency (Anhydrous): ≥845 µg/mg
pH8.0-10.0 (1% solution in water)
Solution TestSoluble (50 mg/ml in water)