4-Bromoaniline, 96%, pract., Thermo Scientific Chemicals
4-Bromoaniline, 96%, pract., Thermo Scientific Chemicals
4-Bromoaniline, 96%, pract., Thermo Scientific Chemicals
4-Bromoaniline, 96%, pract., Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromoaniline, 96%, pract., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ACR402871000
Price (MYR)
601.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
601.00
Add to cart
Chemical Identifiers
CAS1072-62-4
IUPAC Name2-ethyl-1H-imidazole
Molecular FormulaC5H8N2
InChI KeyPQAMFDRRWURCFQ-UHFFFAOYSA-N
SMILESCCC1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay (GC)≥97.5%
Melting Point (clear melt)81.5-87.5?C
Water Content (Karl Fischer Titration)≤1.0%
FormCrystals or powder or crystalline powder or flakes