Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals
CAS: 298-07-7 | C16H35O4P | 322.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass Bottle
Catalog number ACR402470010
Price (MYR)
1,699.00
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,699.00
Add to cart
Chemical Identifiers
CAS93-09-4
IUPAC Namenaphthalene-2-carboxylic acid
Molecular FormulaC11H8O2
InChI KeyUOBYKYZJUGYBDK-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Melting point183°C to 187°C
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Appearance (Form)Powder