Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
CAS: 622-78-6 | C8H7NS | 149.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR402131000
Price (MYR)
1,740.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,740.00
Add to cart
Chemical Identifiers
CAS68-19-9
IUPAC Name(10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-1-cyano-46-hydroxy-12-(hydroxymethyl)-5,6,17,23,28,31,31,36,38,41,42-undecamethyl-15,20-dioxo-11,14,16-trioxa-2λ⁵,9,19,26λ⁵,43,44λ⁵,45λ⁵-heptaaza-15λ⁵-phospha-1-cobaltadodecacyclo[27.14.1.1¹,³⁴.1²,⁹.1¹⁰,¹³.0¹,²⁶.0³,⁸.0²³,²⁷.0²⁵,⁴².0³²,⁴⁴.0³⁹,⁴³.0³⁷,⁴⁵]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate
Molecular FormulaC63H88CoN14O14P
InChI KeyYUWVGNQGDAPKCX-BVWPOUIRNA-L
SMILES[C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Formcrystals or crystalline powder
Assay (unspecified)> 98.0% (on dried material)
Loss on Drying≤ 5.0%
Appearance (Color)Dark red to red-purple