Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
CAS: 622-78-6 | C8H7NS | 149.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR402130250
Price (MYR)
795.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
795.00
Add to cart
Specifications
Chemical Name or MaterialBenzyl isothiocyanate
Name Note98%
CAS622-78-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger