Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzyl isothiocyanate, 98%, Thermo Scientific Chemicals
CAS: 622-78-6 | C8H7NS | 149.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR4021300505 gGlass bottle
Catalog number ACR402130050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzyl isothiocyanate
Name Note98%
CAS622-78-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger