3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific Chemicals
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific Chemicals
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific Chemicals
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
Packaging:
Glass bottle
Catalog number ACR401935000
Price (MYR)
996.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
996.00
Add to cart
Specifications
Chemical Name or Material3,3′,4,4′-Benzophenonetetracarboxylic dianhydride
Name Note99%
CAS2421-28-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning