Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
CAS: 1670-14- | C7H9ClN2 | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR401791000
Price (MYR)
1,199.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,199.00
Add to cart
Specifications
Chemical Name or MaterialBenzamidine hydrochloride
CAS1670-14-
Health Hazard 1GHS Signal Word: Warning
Melting Point167.0°C to 171.0°C