4,4'-Azodianiline, 95%, Thermo Scientific Chemicals
4,4'-Azodianiline, 95%, Thermo Scientific Chemicals
4,4'-Azodianiline, 95%, Thermo Scientific Chemicals
4,4'-Azodianiline, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Azodianiline, 95%, Thermo Scientific Chemicals
CAS: 538-41-0 | C12H12N4 | 212.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR401580050
Price (MYR)
1,836.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,836.00
Add to cart
Specifications
Chemical Name or Material4, 4'-Azodianiline
Name Note95%
CAS538-41-0
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning