Azobenzene, 98%, Thermo Scientific Chemicals
Azobenzene, 98%, Thermo Scientific Chemicals
Azobenzene, 98%, Thermo Scientific Chemicals
Azobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Azobenzene, 98%, Thermo Scientific Chemicals
CAS: 103-33-3 | C12H10N2 | 182.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number ACR401540500
Price (MYR)
1,143.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,143.00
Add to cart
Specifications
Chemical Name or MaterialAzobenzene
CAS103-33-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Suspected of causing genetic defects.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3GHS P Statement:
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.