Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
CAS: 123-99-9 | C9H16O4 | 188.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number ACR401525000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAzelaic Acid
Name Note98%
CAS123-99-9
Melting Point98.0°C to 103.0°C
Boiling Point237.0°C (20.0hPa)