Aluminon, ACS reagent, Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
CAS: 569-58-4 | C22H23N3O9 | 473.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR401501000
Price (MYR)
985.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
985.00
Add to cart
Specifications
Chemical Name or MaterialAluminon
CAS569-58-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiou