Aluminon, ACS reagent, Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
CAS: 569-58-4 | C22H23N3O9 | 473.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR401500250
Price (MYR)
469.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
469.00
Add to cart
Chemical Identifiers
CAS51-67-2
IUPAC Name4-(2-aminoethyl)phenol
Molecular FormulaC8H11NO
InChI KeyDZGWFCGJZKJUFP-UHFFFAOYSA-N
SMILESNCCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)98.0% min.
Melting Point159? to 165?