Anthrone, 95%, pure, Thermo Scientific Chemicals
Anthrone, 95%, pure, Thermo Scientific Chemicals
Anthrone, 95%, pure, Thermo Scientific Chemicals
Anthrone, 95%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Anthrone, 95%, pure, Thermo Scientific Chemicals
CAS: 90-44-8 | C14H10O | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
View all
Glass Bottle
Catalog number ACR401361000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAnthrone
CAS90-44-8
Health Hazard 1GHS Signal Word: Warning
Melting Point152.0°C to 157.0°C
EINECS Number201-994-