2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals
2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals
2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals
2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals
CAS: 121-88-0 | C6H6N2O3 | 154.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number ACR400900500
Price (MYR)
572.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
572.00
Add to cart
Chemical Identifiers
CAS96-53-7
IUPAC Name1,3-thiazolidine-2-thione
Molecular FormulaC3H5NS2
InChI KeyWGJCBBASTRWVJL-UHFFFAOYSA-N
SMILESS=C1NCCS1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point103°C to 106°C
Infrared spectrumConforms
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Titration with TBAH97.5 to 102.0 %