1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
CAS: 140-31-8 | C6H15N3 | 129.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR400765000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS84-58-2
IUPAC Name4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
Molecular FormulaC8Cl2N2O2
InChI KeyHZNVUJQVZSTENZ-UHFFFAOYSA-N
SMILESClC1=C(Cl)C(=O)C(C#N)=C(C#N)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
FormPowder or crystalline powder or crystals
Assay (Iodometric Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Melting Point (clear melt)213.0-219.0°C