1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
CAS: 140-31-8 | C6H15N3 | 129.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR400761000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1317-38-0
IUPAC Namecopper(2+) oxidandiide
Molecular FormulaCuO
InChI KeyKKCXRELNMOYFLS-UHFFFAOYSA-N
SMILES[O--].[Cu++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.3% max.