Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ACR400180500
Price (MYR)
3,649.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
3,649.00
Add to cart
Chemical Identifiers
CAS4282-40-0
IUPAC Name1-iodoheptane
Molecular FormulaC7H15I
InChI KeyLMHCYRULPLGEEZ-UHFFFAOYSA-N
SMILESCCCCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
CommentCopper powder used as stabilizer
Refractive Index1.4880-1.4920 @ 20?C
Appearance (Color)Clear colorless to pale pink
Assay (GC)>97.5%