Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number ACR400180100
Price (MYR)
994.00
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
994.00
Add to cart
Chemical Identifiers
CAS5970-45-6
IUPAC Namezinc(2+) diacetate dihydrate
Molecular FormulaC4H10O6Zn
InChI KeyBEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILESO.O.[Zn++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
FormCrystals or powder or crystalline powder or fused solid
Complexometric Titration≥97.0 to ≤103.0%