1'-Acetonaphthone, 95%, Thermo Scientific Chemicals
1'-Acetonaphthone, 95%, Thermo Scientific Chemicals
1'-Acetonaphthone, 95%, Thermo Scientific Chemicals
1'-Acetonaphthone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1'-Acetonaphthone, 95%, Thermo Scientific Chemicals
CAS: 941-98-0 | C12H10O | 170.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number ACR400092500
Price (MYR)
839.00
BOT
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
839.00
BOT
Add to cart
Specifications
Chemical Name or Material1'-Acetonaphthone
Name Note95%
CAS941-98-0
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning