3-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals
3-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals
3-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals
3-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals
CAS: 85-38-1 | C7H5NO5 | 183.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR399820010
Price (MYR)
486.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
486.00
Add to cart
Chemical Identifiers
CAS2269-22-9
IUPAC Namealuminium(3+) tris(butan-2-olate)
Molecular FormulaC12H27AlO3
InChI KeyWOZZOSDBXABUFO-UHFFFAOYNA-N
SMILES[Al+3].CCC(C)[O-].CCC(C)[O-].CCC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly hazy colorless to light yellow
Infrared spectrumConforms
Assay>=96.0 % (ex Al)
Refractive index1.4380 to 1.4400 (20°C, 589 nm)
Appearance (Form)Viscous liquid