Check Order Status
2-Chloro-3-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™
Element Search
Structure Search
2-Chloro-3-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™
Have Questions?
Catalog Number
Quantity
Packaging
ACR398430010
1 g
Glass bottle
Catalog number
ACR398430010
Price (MYR)
126.03
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
126.03
Add to cart
Specifications
Chemical Name or Material
2-chloro-3-(trifluoromethyl)benzaldehyde
CAS
93118-03-7
CAS Min %
97.5
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0