Glass beads, 400-800 micron, Thermo Scientific Chemicals
Glass beads, 400-800 micron, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glass beads, 400-800 micron, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ACR397645000
Price (MYR)
1,406.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,406.00
Add to cart
Chemical Identifiers
CAS623-50-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
Refractive index1.4170 to 1.4200 (20°C, 589 nm)