Glass beads, 400-800 micron, Thermo Scientific Chemicals
Glass beads, 400-800 micron, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glass beads, 400-800 micron, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number ACR397641000
Price (MYR)
596.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
596.00
Add to cart
Chemical Identifiers
CAS71-23-8
IUPAC Namepropan-1-ol
Molecular FormulaC3H8O
InChI KeyBDERNNFJNOPAEC-UHFFFAOYSA-N
SMILESCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on Evaporation4ppm
Residual water0.08%