Glass beads, 400-800 micron, Thermo Scientific Chemicals
Glass beads, 400-800 micron, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glass beads, 400-800 micron, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number ACR397640250
Price (MYR)
436.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
436.00
Add to cart
Chemical Identifiers
CAS360576-04-1
IUPAC Name6-bromo-2,3-difluorobenzaldehyde
Molecular FormulaC7H3BrF2O
InChI KeyLAVPYRPTHABUAD-UHFFFAOYSA-N
SMILESFC1=C(F)C(C=O)=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused/lumpy solid
Free acid (titration)≤1.5%
Melting Point (clear melt)38.5-44.5?C
Appearance (Color)White to cream or pale brown
Assay (GC)≥97.5%