Methyl sulfoxide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number ACR397600010
Price (MYR)
1,330.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
1,330.00
Add to cart
Chemical Identifiers
CAS119-91-5
IUPAC Name2,2'-biquinoline
Molecular FormulaC18H12N2
InChI KeyWPTCSQBWLUUYDV-UHFFFAOYSA-N
SMILESC1=CC2=CC=C(N=C2C=C1)C1=NC2=CC=CC=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or flakes
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)192.0-199.0?C
Appearance (Color)White to cream to pale brown