4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
CAS: 73870-24-3 | C6H7Br2N | 252.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ACR397330250
Price (MYR)
1,821.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,821.00
Add to cart
Chemical Identifiers
CAS50926-11-9
IUPAC Namestannanone indium
Molecular FormulaInOSn
InChI KeyAMGQUBHHOARCQH-UHFFFAOYSA-N
SMILES[In].O=[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.