4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
CAS: 73870-24-3 | C6H7Br2N | 252.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ACR397330050
Price (MYR)
593.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
593.00
Add to cart
Chemical Identifiers
CAS9001-73-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light brown
Infrared spectrumConforms
Assay>=1000 TU/mg (Enz.act.) accurancy:+/-5%
Loss on drying=<7.0 % (60°C, 4 hrs) (vacuum)
pH4.8 to 6.2 (1 in 50 solution)