Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
2.5 kg
Catalog number ACR397282500
Price (MYR)
567.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
567.00
Add to cart
Chemical Identifiers
CAS9003-69-4
MDL NumberMFCD00132704
SpecificationsSpecification SheetSpecification Sheet
Moisture holding capacity54.0 to 60.0 %
Uniformity coefficient=<2.0
Particle size=<5.0 % (< 0.300 mm)
Appearance (Color)White to yellow
Appearance (Form)Beads