Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
500 g
2.5 kg
Catalog number ACR397280010
Price (MYR)
1,136.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
1,136.00
Add to cart
Chemical Identifiers
CAS30525-89-4
IUPAC Nameformaldehyde
Molecular FormulaCH2O
InChI KeyWSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILESC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or beads
Infrared spectrumConforms
Titration with HCl>=95 %
Ash=<0.02 %