Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Catalog number ACR396982500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS16650-55-8
IUPAC Name4-bromonaphthalene-1-carboxylic acid
Molecular FormulaC11H7BrO2
InChI KeyFIJIPZQZVLCOMB-UHFFFAOYSA-N
SMILESOC(=O)C1=C2C=CC=CC2=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%