Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Catalog number ACR396980025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS617-84-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4330 to 1.4350 (20°C, 589 nm)