Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, specified according to the requirements of Ph. Eur., Thermo Scientific Chemicals
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
2.5 L
Catalog number ACR396980010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7367-88-6
IUPAC Nameethyl (2E)-dec-2-enoate
Molecular FormulaC12H22O2
InChI KeyGNJARWZWODMTDR-ZHACJKMWSA-N
SMILESCCCCCCCC=CC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4430-1.4480 @ 20?C