2-Butanone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
2-Butanone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
2-Butanone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
2-Butanone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
2-Butanone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Butanone, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
CAS: 78-93-3 | C4H8O | 72.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR396951000100 mLAcroSeal™ Glass Bottle
Catalog number ACR396951000
Price (MYR)
603.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
603.00
Add to cart
Chemical Identifiers
CAS121-57-3
IUPAC Name4-aminobenzene-1-sulfonic acid
Molecular FormulaC6H7NO3S
InChI KeyHVBSAKJJOYLTQU-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after ignition=<0.01 %
Appearance (Color)Off-white
Infrared spectrumConforms
Insoluble matter=<0.02 % (in sodium carbonate solution)
Nitrite (NO2)=<0.5 ppm