5-Aza-2'-deoxycytidine, 98%, Thermo Scientific Chemicals
5-Aza-2'-deoxycytidine, 98%, Thermo Scientific Chemicals
5-Aza-2'-deoxycytidine, 98%, Thermo Scientific Chemicals
5-Aza-2'-deoxycytidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Aza-2'-deoxycytidine, 98%, Thermo Scientific Chemicals
CAS: 2353-33-5 | C9H13N3O4 | 227.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
Packaging:
Glass bottle
Catalog number ACR396810100
Price (MYR)
810.00
Add to cart
Quantity:
10 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
810.00
Add to cart
Specifications
Chemical Name or Material5-Aza-2′-deoxycytidine
CAS2353-33-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0