Di(ethylene glycol) divinyl ether, 95%
Di(ethylene glycol) divinyl ether, 95%
Di(ethylene glycol) divinyl ether, 95%
Di(ethylene glycol) divinyl ether, 95%
Thermo Scientific Chemicals
Di(ethylene glycol) divinyl ether, 95%
CAS: 764-99-8 | C8H14O3 | 158.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR39541050050 mLGlass bottle
Catalog number ACR395410500
Price (MYR)
1,605.00
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,605.00
Add to cart
Chemical Identifiers
CAS7558-79-4
IUPAC Namedisodium hydrogen phosphate
Molecular FormulaHNa2O4P
InChI KeyBNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<20 ppm
Iron (Fe)=<20 ppm
Sulfate (SO4)=<50 ppm
Appearance (Color)White
Appearance (Form)Powder or granular powder