Quartz Wool
Thermo Scientific Chemicals
Quartz Wool
Filter aid
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Catalog number ACR393610100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS100-51-6
IUPAC Namephenylmethanol
Molecular FormulaC7H8O
InChI KeyWVDDGKGOMKODPV-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)20, max
Impurity content0.02%, max, as Acetophenone C₆H₅COCH₃ ( by GC)
Assay (GC)≥99.0%
Impurity content0.01%, max, as Benzaldehyde C₆H₅CHO ( by GC)
FormClear liquid