Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Plastic bottle
Catalog number ACR393320010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1310-58-3, 64-17-5
Specifications
Chemical Name or MaterialPotassium hydroxide
ColorBrown-Orange to Yellow
Name Note1N solution in ethanol
Boiling Point78.0°C
CAS1310-58-3
View more
Suitable for battery materials development.
For Manufacturing and Laboratory Use. Not for beverage use