1,3-Dimethylbarbituric acid, 98%
Product Image
Thermo Scientific Chemicals
1,3-Dimethylbarbituric acid, 98%
CAS: 769-42-6 | C6H8N2O3 | 156.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR393285000500 gGlass Bottle
Catalog number ACR393285000
Price (MYR)
1,749.00
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,749.00
Add to cart
Chemical Identifiers
CAS3326-71-4
IUPAC Namefuran-2-carbohydrazide
Molecular FormulaC5H6N2O2
InChI KeySKTSVWWOAIAIKI-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Melting point76°C to 80°C
Appearance (Color)Beige
GC>=97.5 %
Infrared spectrumConforms
  • 1,3-Dimethylbarbituric acid, 98% 500g Plastic Bottle Cas No. 769-42-6
RUO – Research Use Only