2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals
CAS: 7305-71-7 | C4H6N2S | 114.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Catalog number ACR392790250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS375-85-9
IUPAC Nametridecafluoroheptanoic acid amine
Molecular FormulaC7H4F13NO2
InChI KeyJVZREVRTMWNFME-UHFFFAOYSA-N
SMILESN.OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or fused solid or clear liquid as melt
Identification (FTIR)Conforms (UK sourced material only)
Appearance (Color)White to pale yellow/orange
Assay from Supplier's CofA≥98.0 to ≤102.0% (Titration, US sourced material only)
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0% (UK sourced material only)