2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, For Grignard reaction, anhydrous, stabilized, ACROS Organics, Quantity: 10L, Packaging: Metal Drum, Boiling Point: 78.0 deg.C to 80.0 deg.C, CAS: 96-47-9, Colorless, Grade: For Grignard reaction, Melting Point: -136.0 deg.C, Molecular Formula: C5H10O, Molecular Weight: 86.13
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number ACR392380100
Price (MYR)
5,445.00
EA
Add to cart
Quantity:
10 L
Packaging:
Metal Drum
Request bulk or custom format
Price (MYR)
5,445.00
EA
Add to cart
Chemical Identifiers
CAS779-02-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (GC)≥98.5%
Melting Point (clear melt)75-82°C